Learn More
Greiner Bio-One Masterblock™ 96-well MicroPlates
Product Code. 10023241
Change view
Click to view available options
Well Shape:
Conical Bottom (V-well)
Round Bottom (U-well)
Total Volume (Metric) Well:
1 mL
2 mL
Unit Size:
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Well Shape Total Volume (Metric) Well Unit Size
10023241 Round Bottom (U-well) 1 mL Pack of 50
10688821 Conical Bottom (V-well) 2 mL Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10023241 Supplier Greiner Bio-One Supplier No. 780201
Sample Preparation and Storage Essentials
Use promo code 27996
View all promotions
Estimated Shipment: 15-10-2026
Manufactured from polypropylene for excellent chemical and thermal resistance. Greiner Bio One™ Masterblock™ 96-Well Microplates feature raised wells to accommodate thermal or adhesive sealing.
Unique Masterblock design is suitable for applications where larger working volumes are required such as liquid storage in compound libraries.
  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • Manufactured under DIN ISO 9001 guidelines
  • Traceable to production through a defined LOT number system
  • Ideal microplates for storing sample material and cultivating bacteria or yeasts
  • Compatible with automated systems
  • Uniform external dimensions and tolerances
  • Alphanumeric well coding
  • High chemical resistance
  • High temperature resistance (-196°C to + 121°C)
  • Sealable with adhesive films and heat sealer 
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance DIN ISO 9001
Certified Yes
Format Standard
Lid Without Lid
Array 8 x 12
Color Clear
Material Polypropylene
No. of Wells 96
Sterility Non-sterile
Surface Treatment Non-treated
Total Volume (Metric) Well 1 mL
Well Shape Round Bottom (U-well)
Dnase Free Yes
For Use With (Application) Ideal microplates for storing sample material but are also ideal for cultivating bacteria or yeasts
Plate Compatibility Yes
Quantity 50/Cs.
RNase Free Yes
Temperature Range (Metric) –196 to 121°C