Learn More
Atago™ MASTER-S/Millα Salinity Refractometer
Description
- Durable against salty samples
- Displays salinity in parts per million
- Automatic temperature compensation
- Water resistant
Specifications
Specifications
| Type | Salinity Refractometer |
| Scale Range | 0 to 100 Salinity, 1.000 to 1.070 Specific Gravity |
| Model | MASTER-S/Millα |
| Accuracy | ±2‰ Salinity, ±0.001 Specific gravity at 10 to 30°C |
| Repeatability | ±1‰ Salinity, ±0.0005 Specific gravity |
| Scale Type | Salinity, Specific gravity |
| For Use With (Application) | Measuring the salinity and specific gravity of seawater |
| IP Rating | IP65 |
| Weight (Metric) | 110 g |
| Dimensions (L x W x H) | 3.2 x 3.4 x 20.7 cm |