Learn More
MARK1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21552-1-AP-150UL
Description
The MARK1 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human MARK1. This antibody recognizes human, mouse, rat antigen. The MARK1 antibody has been validated for the following applications: IHC, IP, WB, ELISA analysis.
Specifications
| MARK1 | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 4139 | |
| IgG |
| Western Blot, Immunoprecipitation, Immunohistochemistry, ELISA | |
| 360 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Rat, Mouse |