Learn More
Cole-Parmer™ Mark-10 Digital Torque Gauge
Product Code. 15524999 Shop All Cole-Parmer Products
Change view
Click to view available options
Accuracy:
±0.5%
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Accuracy Unit Size
15524999 ±0.5% Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15524999 Supplier Cole-Parmer™ Supplier No. 2530164
Estimated Shipment: 22-12-2026
Offers quality control solutions for a variety of industries including manufacturing, plastics and consumer products for torque testing up to 100 pounds per inch. Cole-Parmer™ Mark-10 Digital Torque Gauge provides precision accuracy for clockwise and counter-clockwise torque testing.
  • The easy push-button operation allows you to easily select units of measurement, reset zero and recall peak loads
  • Torque gauge can be powered by either rechargeable battery or AC adapter
  • Rugged construction allows gauge to be used by hand or mounted to a test stand
  • Order manual or motorized test stand separately
  • Peaked torque values saved and displayed
  • Programmable set points with pass/fail indicators
  • Includes carrying case, AC adapter, rechargeable battery, chuck key, USB cable, MESURLite software and MESURgauge demo software
Specifications
Height (English) 3.875 in.
Includes Carrying case, AC adapter, rechargeable battery, chuck key, USB cable, MESURLite software and MESURgauge demo software.
Width (English) 2.5 in.
Accuracy ±0.5%
Product Type Mark-10 Digital Torque Gauge