Learn More
SCAT™ Safety Funnel 'MARCO' with Lid, V2.0
Product Code. p-7203079 Shop All SCAT Europe Products
Click to view available options
Includes:
Funnel w/ Hinged Lid, No Lance
Height (Metric):
150 mm
Unit Size:
Each
Stopper Number:
S50
S55
S60/61
S65
Product Code. 16609095
Brand: SCAT™ 318952
Only null left
Add to basket
Add to basket
The 'MARCO' safety funnel can be safely closed and prevent danger of ignition, and is also available in conductive HDPE-EC material.
  • Modern lid funnel made of high-quality HDPE with very high resistance to acids, alkalis and many chemicals
  • Integrated gripping edge on the funnel lid increases the grip without having to touch the possibly contoured lid
  • Innovative funnel lid with integrated sealing ring closes the funnel edge and prevents the escape of explosive or health-endangering gases
  • Sturdy hinge guarantees a long service life of the closing mechanism
  • Rotating screw cap makes it easier to screw the lid funnel on and off the canister
  • Removable sieve for easy cleaning
  • Non-conductive version of the MARCO V2.0 is in line with the SCAT color scheme due to its blue and white coloring
  • Conductive versions of the MARCO V2.0 are in black and thus clearly differ in color from the non-conductive version
Specifications
Diameter (Metric)Top 140 mm
Capacity (Metric) 300 mL
Material HDPE
Height (Metric) 150 mm
Autoclavable Not Autoclavable
Color Blue, White
Includes Funnel w/ Hinged Lid, No Lance
Product Type Safety Funnel