Learn More
Emma Safety Footwear Marco (Frontier 112) Safety Shoes
Product Code. 15998181
Change view
Click to view available options
Size:
39
40
41
42
43
44
46
47
48
Unit Size:
1 pair
Product Code. Size unitSize
15998181 46 1 pair
15978181 39 1 pair
15988181 40 1 pair
15612469 41 1 pair
15775233 42 1 pair
15865771 43 1 pair
15785233 44 1 pair
15908191 47 1 pair
15918191 48 1 pair
9 options
Product Code. 15998181
Brand: Emma Safety Footwear MM11209046
Only null left
Add to basket
Add to basket
Made with the same care and  attention to detail associated with  high end luxury shoes. Brazilian 2 tone full grain leather combined with nubuck adds to the stylish look.
  • S3-Safety shoe with sole plate for wet work
  • Full grain leather construction
  • Thermoplastic Polyurethane Outsole
  • ESD certified
  • Chrome-free lining
  • Sanitized
Specifications
Certifications/Compliance EN ISO 20345:2011, EN 12568:2010
Closure Type Laces
Color Brown
Design Marco (Frontier 112)
Liner Material Hydro-Tec™ Sanitized Silver & Wellness leather
Material Leather
Size 46
Toe Material Steel