Learn More
MAPK7 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
Antigen MAPK7
Concentration 0.3 mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16886183
View Documents
Proteintech
10036-2-AP-20UL
20 μL
1710.33 NOK
20µL
View alternative products
Save Up to 
»
16876183
View Documents
Proteintech
10036-2-AP-150UL
150 μL
4836.10 NOK
150µL
View alternative products
Save Up to 
»
Description
Description
MEK5 and ERK5, a member of the MAP kinase subfamily of the Ser/Thr protein kinases, interact specifically with one another and not with MEK1/ERK1 or MEK2/ERK2 pathways. ERK5 is activated by tyrosine and threonine phosphorylation It is expressed in many adult tissues, abundantly in heart, placenta, lung, kidney and skeletal muscle, but is not detectable in liver. The second proline-rich region may interact with actin targeting the kinase to a specific location in the cell. ERK5 is autophosphorylated on threonine and tyrosine residues when the C-terminal part of the kinase, which could have a regulatory role, is absent.
Specifications
Specifications
MAPK7
Western Blot
Unconjugated
Rabbit
Mouse, Human
Q13164, Q9WVS8
23939, 5598
MAPK7 Fusion Protein Ag0017
Primary
-20°C
MAPK7
0.3 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Big MAP kinase 1, BMK 1, BMK1, ERK 5, ERK4, ERK5, MAP kinase 7, MAPK 7, MAPK7, PRKM7
MAPK7
IgG
Antigen Affinity Chromatography