missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAPK15 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13452-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The ERKs are a subfamily of the MAPKs that have been implicated in cell growth and differentiation. Extracellular signal-regulated kinase 8 (Erk8) is a large MAP kinase whose activity is controlled by serum and the c-Src non-receptor tyrosine kinase. ERK8 down-regulates transactivation of the glucocorticoid receptor through Hic-5 and can negatively regulate transcriptional co-activation of androgen receptor and GRalpha by Hic-5 in a kinase-independent manner, suggesting a broader role for ERK8 in the regulation of nuclear receptors beyond estrogen receptor alpha. Erk8 is a novel effector of RET/PTC3 and, therefore, RET biological functions.Specifications
MAPK15 | |
Polyclonal | |
Unconjugated | |
MAPK15 | |
ERK 7, ERK 8, ERK7, ERK8, MAP kinase 15, MAPK 15, MAPK15 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
225689, 286997, 332110 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q80Y86, Q8TD08, Q9Z2A6 | |
Mapk15 | |
MAPK15 Fusion Protein Ag4255 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |