Learn More
Innopsys MAPIX Analyzer
Description
- For microarray image analysis and data quantification
- Intuitive and straightforward handling using .GAL files
- Available for Windows 32 and 64 bit operating systems
Specifications
Specifications
| Type | Software |
| For Use With (Application) | Microarray Image Analysis and Data Quantification |
| For Use With (Equipment) | Windows Operating Systems (32/64 Bit) |
| Includes | Full Single User License |