Learn More
MAP4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Product Code. 16848163 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16848163 150 μL 150µL
16858163 20 μL 20µL
2 options
Product Code. 16848163
Brand: Proteintech 112291AP150UL
Rabbit Polyclonal Antibody
This gene belongs to the forkhead family of transcription factors which is characterized by a distinct DNA-binding forkhead domain. The specific function of this gene has not yet been determined; however, it has been shown to play a role in the regulation of embryonic and ocular development. Mutations in this gene cause various glaucoma phenotypes including primary congenital glaucoma, autosomal domit iridogoniodysgenesis anomaly, and Axenfeld-Rieger anomaly.
Specifications
Antigen MAP4
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene MAP4
Gene Accession No. P27546, P27816, Q5M7W5
Gene Alias MAP 4, MAP4
Gene Symbols Map4
Host Species Rabbit
Immunogen MAP4 Fusion Protein Ag1741
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17758, 367171, 4134
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG