missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MAP3K14 (NIK), GST Tag Recombinant Protein

Human MAP3K14 (NIK), GST Tag Recombinant Protein

Brand:  Invitrogen™ PV4902

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10307993

  • 5110.00 NOK / Each
Estimated Shipment: 27-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

MAP3K14 (NIK) is structurally related to MAP3Ks. MAP3K14 phosphorylates and activates CHUK (IKKalpha) and IKBKB (IKK beta), which in turn phosphorylate IkB-alpha, releasing NF-KappaB leading to up-regulation of inflammatory response genes.
Specifications

Specifications

NP_003945.2
9020
10 μg
STE Unique Kinase Family
Human
GST Tag
Biochemical binding/kinetic
Dry Ice
mitogen-activated protein kinase kinase kinase 14
Kinases & Inhibitors
Liquid
Signal Transduction
Kinase Assay
96.9 kDa
MAP3K14 (NIK)
aa 318-947
Recombinant
Storage Buffer: 50 mM Tris pH 7.5, 150 mM NaCl, 0.02% Trition X-100, 0.5 mM EDTA, 2 mM DTT, 50% Glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Ser⁄Thr Kinases: STE Group
MAP3K14
Baculovirus
Recombinant, Catalytic
Serine/Threonine Kinases
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers