Learn More
MAP1B Mouse anti-Human, Unconjugated, Clone: 1E6B9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67423-1-IG-20UL
This item is not returnable.
View return policy
Description
The MAP1B antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human MAP1B. This antibody recognizes human, rat, mouse antigen. The MAP1B antibody has been validated for the following applications: IF, IHC, ELISA analysis.
Specifications
| MAP1B | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat |
| Immunohistochemistry, Immunofluorescence, ELISA | |
| 1E6B9 | |
| Unconjugated | |
| 20 μL | |
| 4131 | |
| IgG1 |