Learn More
Proteintech MAP1B Mouse anti-Human, Unconjugated, Clone: 1E6B9, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67423-1-IG-20UL
Additional Details : Weight : 0.00020kg
 View more versions of this product
Product Code. 17521426
  • 1415.13 NOK / 20µL
Estimated Shipment: 06-06-2023
Description
The MAP1B antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human MAP1B. This antibody recognizes human, rat, mouse antigen. The MAP1B antibody has been validated for the following applications: IF, IHC, ELISA analysis.
Specifications
Specifications
MAP1B
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat
Immunohistochemistry, Immunofluorescence, ELISA
1E6B9
Unconjugated
20 μL
4131
IgG1