Learn More
SI Analytics™ Cannon-Fenske Reverse Flow Viscometer
Product Code. 10707021 Shop All SI Analytics Products
Change view
Click to view available options
Viscosity Range:
20 to 100 cSt
3 to 15 cSt
7 to 35 cSt
Approximate Constant:
0.0215K
0.035K
0.1K
Size:
0.63mm Diameter
0.78mm Diameter
1.02mm Diameter
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Viscosity Range Approximate Constant Size Unit Size
10707021 3 to 15 cSt 0.0215K 0.63mm Diameter Each
10538981 7 to 35 cSt 0.035K 0.78mm Diameter Each
10423711 20 to 100 cSt 0.1K 1.02mm Diameter Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10707021 Supplier SI Analytics™ Supplier No. 285403034
  • ONLINE EXCLUSIVE PRICE
    5485.00 NOK
    4582.89 NOK / Pack of 1
    Save 902.11 NOK
    16% Off
Estimated Shipment: 17-12-2026
Determine absolute and relative kinematic viscosity of liquids with Newtonian flow behavior. SI Analytics™ Cannon-Fenske Reverse Flow Viscometer includes three ring marks since they can only be checked by means of manual measurements.
  • Suitable for transparent liquids manual measurement, opaque liquids manual measurement, foaming liquids, liquid mixture with highly volatile components, and high or low temperature measurements
  • Calibrated
  • With two constants
  • With three ring marks
  • Only for manual measurements
  • Filling quantity of 12mL
  • Overall length of 295mm
TRUSTED_SUSTAINABILITY
Specifications
Product Type Viscometer
Viscosity Range 3 to 15 cSt
Calibrated Yes
Description Capillary No. 100
Approximate Constant 0.0215K
ASTM Numbers D 446
Certifications/Compliance ISO/DIS 3105, ASTMD 2515, ASTM D 446, NF T 60 - 100
Includes Ring Marks and 2 Constants
For Use With (Application) Only for manual measurement
Size 0.63mm Diameter
Volume (Metric) 12 mL