Learn More
SI Analytics™ Manual Ubbelohde Viscometer
Product Code. 12068163 Shop All SI Analytics Products
Change view
Click to view available options
Approximate Constant:
0.001K
0.003K
0.01K
0.03K
0.05K
0.1K
0.3K
1K
3K
Size:
0.36mm Capillary Diameter
0.47mm Capillary Diameter
0.63mm Capillary Diameter
0.84mm Capillary Diameter
0.95mm Capillary Diameter
1.13mm Capillary Diameter
1.50mm Capillary Diameter
2.01mm Capillary Diameter
2.65mm Capillary Diameter
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Approximate Constant Size Unit Size
12068163 0.3K 1.50mm Capillary Diameter Each
10150181 0.001K 0.36mm Capillary Diameter Each
10231881 0.003K 0.47mm Capillary Diameter Each
12018163 0.01K 0.63mm Capillary Diameter Each
12038163 0.03K 0.84mm Capillary Diameter Each
12048163 0.1K 1.13mm Capillary Diameter Each
12078163 1K 2.01mm Capillary Diameter Each
12088163 3K 2.65mm Capillary Diameter Each
12028163 0.05K 0.95mm Capillary Diameter Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 12068163 Supplier SI Analytics™ Supplier No. 285400078
  • ONLINE EXCLUSIVE PRICE
    6070.00 NOK
    5454.83 NOK / Pack of 1
    Save 615.17 NOK
    10% Off
View all promotions
 Restricted Item
Estimated Shipment: 18-12-2026
Determine absolute and relative kinematic viscosity of liquids with Newtonian flow behavior. SI Analytics™ Manual Ubbelohde Viscometer includes ring marks to ensure that they can be checked by means of manual measurements.
  • Suitable for transparent liquids manual and automatic measurement, foaming liquids, liquid mixture with highly volatile components, and high or low temperature measurements
  • The recommended minimum flow through time is 200 seconds
  • Calibrated with constant for manual measurements
  • Filling quantity of 15 to 20mL
TRUSTED_SUSTAINABILITY
Specifications
Product Type Viscometer
Calibrated Yes
Description 290mm Long
Approximate Constant 0.3K
Certifications/Compliance DIN 51 562 Part 1, ISO/DIS 3105 (BS-IP-SL)
Includes Ring Marks
For Use With (Application) For Determination of Absolute and Relative Kinematic Viscosity of Liquids With Newtonian Flow Behavior
Size 1.50mm Capillary Diameter
Volume (Metric) 15 to 25 mL