Learn More
Hamilton™ Manual HPLC Cemented Needle Syringes
Product Code. p-7138549
Click to view available options
Connection Type:
Cemented
Luer Tip, Cemented
Unit Size:
Each
Volume (Metric):
250 μL
Product Code. 11598044
Brand: Hamilton™ DS80365
Specifically designed for manual or automatic injection into high pressure ports with Rheodyne™, Valco™, Beckman™ and SSI™ valves.
  • Needle is blunt and electro-polished to slide smoothly into the injection port
  • Available as standard cemented needle or Luer tip cemented needle
  • 51mm in length with point style 3
Specifications
Needle Gauge 22s G
Length (Metric) Needle 51 mm
Needle Point Style 3
Connection Type Cemented