Learn More
Fisherbrand™ Manual Decapper, Steel, Non-autoclavable
Shop All Fisherbrand Products
Click to view available options
Size:
8 mm
11 mm
13 mm
20 mm
28 mm
32 mm
Unit Size:
Each
Description
- Decapper
- Manual
- Steel
Specifications
Specifications
| Operation | Manual |
| Type | Manual Decapper |
| For Use With (Equipment) | Aluminum Cap |
| Size | 28 mm |