Learn More
Thermo Scientific™ Manual Crimpers, Decrimpers and Decrimping Pliers
Product Code. 15188784
Change view
Click to view available options
Size:
8 mm
11 mm
13 mm
20 mm
28 mm
30 mm
32 mm
Unit Size:
Each
Type:
Manual Crimper
Manual Crimper (Flip-off)
Manual Decapping Plier
Manual Decrimper
Product Code. Size unitSize Type
15188784 20 mm Each -
15280389 8 mm Each Manual Crimper
14860742 11 mm Each Manual Crimper
13488988 20 mm Each Manual Crimper
10147664 8 mm Each Manual Crimper
10429454 11 mm Each Manual Crimper
10566765 13 mm Each Manual Crimper
10231585 13 mm Each Manual Crimper (Flip-off)
12400268 20 mm Each Manual Crimper
10372525 20 mm Each Manual Crimper (Flip-off)
10250535 28 mm Each Manual Crimper
10773667 30 mm Each Manual Crimper
10229964 32 mm Each Manual Crimper
11528470 11 mm Each Manual Crimper
11538470 20 mm Each Manual Crimper
15290389 8 mm Each -
10259614 8 mm Each Manual Decrimper
10488084 11 mm Each Manual Decrimper
10240535 13 mm Each Manual Decrimper
10279364 20 mm Each Manual Decrimper
12852344 28 mm Each Manual Decrimper
10301535 30 mm Each Manual Decrimper
11528240 32 mm Each Manual Decrimper
11564815 20 mm Each Manual Crimper
10530303 11 mm Each Manual Decrimper
10143362 20 mm Each Manual Decrimper
12054907 11 mm Each Manual Decapping Plier
10299454 13 mm Each Manual Decapping Plier
10066164 20 mm Each Manual Decapping Plier
10200205 30 mm Each Manual Decapping Plier
30 options
Product Code. 15188784
Brand: Thermo Scientific™ DCB20C
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Achieve secure, repeatable crimps or remove seals with minimized risk of damage.
Achieve secure, repeatable crimps or remove seals with minimized risk of damage when you use Thermo Scientific™ Manual Crimpers, Decrimpers and Decrimping Pliers, also for use with National or Chromacol vials.
Manual Crimpers
Lightweight, precision crimpers attach seals to chromatography or serum vials. Available for standard aluminum or flip-off seals.
  • Factory set to properly crimp standard-thickness seals
  • Adjustable handle setting screw allows proper crimps with septa of varying thickness
Manual Decrimpers
Forged pliers feature a fitted gripping surface designed to allow removal of the listed sizes of seals.
  • Simply grip pliers around top of seal, squeeze and pry upward to remove
Manual Decrimping Pliers
Place decrimping head squarely on seal and squeeze.
  • Minimizes damage to vial rim with chromatography or serum vials
  • Removes standard aluminum seals and many steel seals
TRUSTED_SUSTAINABILITY
Specifications
Description Manual Decrimper
No. per Pack 1 Ea.
For Use With (Equipment) 20 mm Standard Seal, National Vial, Chromacol Vial
Size 20 mm
For Use With (Application) Industrial Chromatography
Product Line Chromacol