Learn More
- Home
- Products
- Vials
- Vial Capping and Decapping Equipment
- Thermo Scientific™ Manual Crimpers, Decrimpers and Decrimping Pliers
Thermo Scientific™ Manual Crimpers, Decrimpers and Decrimping Pliers
Product Code. 10299454| Product Code. | Size | Type | unitSize |
|---|---|---|---|
| 10299454 | 13 mm | Manual Decapping Plier | Each |
| 15280389 | 8 mm | Manual Crimper | Each |
| 14860742 | 11 mm | Manual Crimper | Each |
| 13488988 | 20 mm | Manual Crimper | Each |
| 10147664 | 8 mm | Manual Crimper | Each |
| 10429454 | 11 mm | Manual Crimper | Each |
| 10566765 | 13 mm | Manual Crimper | Each |
| 10231585 | 13 mm | Manual Crimper (Flip-off) | Each |
| 12400268 | 20 mm | Manual Crimper | Each |
| 10372525 | 20 mm | Manual Crimper (Flip-off) | Each |
| 10250535 | 28 mm | Manual Crimper | Each |
| 10773667 | 30 mm | Manual Crimper | Each |
| 10229964 | 32 mm | Manual Crimper | Each |
| 11528470 | 11 mm | Manual Crimper | Each |
| 11538470 | 20 mm | Manual Crimper | Each |
| 15290389 | 8 mm | - | Each |
| 15188784 | 20 mm | - | Each |
| 10259614 | 8 mm | Manual Decrimper | Each |
| 10488084 | 11 mm | Manual Decrimper | Each |
| 10240535 | 13 mm | Manual Decrimper | Each |
| 10279364 | 20 mm | Manual Decrimper | Each |
| 12852344 | 28 mm | Manual Decrimper | Each |
| 10301535 | 30 mm | Manual Decrimper | Each |
| 11528240 | 32 mm | Manual Decrimper | Each |
| 11564815 | 20 mm | Manual Crimper | Each |
| 10530303 | 11 mm | Manual Decrimper | Each |
| 10143362 | 20 mm | Manual Decrimper | Each |
| 12054907 | 11 mm | Manual Decapping Plier | Each |
| 10066164 | 20 mm | Manual Decapping Plier | Each |
| 10200205 | 30 mm | Manual Decapping Plier | Each |
Product Code. 10299454
Brand: Thermo Scientific™ C4013101
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Description
Achieve secure, repeatable crimps or remove seals with minimized risk of damage when you use Thermo Scientific™ Manual Crimpers, Decrimpers and Decrimping Pliers, also for use with National or Chromacol vials.
Lightweight, precision crimpers attach seals to chromatography or serum vials. Available for standard aluminum or flip-off seals.
- Factory set to properly crimp standard-thickness seals
- Adjustable handle setting screw allows proper crimps with septa of varying thickness
Manual Decripmers
Forged pliers feature a fitted gripping surface designed to allow removal of the listed sizes of seals.
- Simply grip pliers around top of seal, squeeze and pry upward to remove
Manual Decrimping Pliers
Place decrimping head squarely on seal and squeeze.
- Minimizes damage to vial rim with chromatography or serum vials
- Removes standard aluminum seals and many steel seals
Specifications
Specifications
| Description | Decapping Pliers |
| No. per Pack | 1 Ea. |
| Type | Manual Decapping Plier |
| For Use With (Equipment) | National Vial, Chromacol Vial |
| Size | 13 mm |
| For Use With (Application) | Industrial Chromatography |
| Product Line | National |