Learn More
Scotsman™ Ice Flaker with Storage Bin
Product Code. 12338697
Change view
Click to view available options
Capacity (Metric):
25 kg
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
12338697 25 kg Each
1 options
Product Code. 12338697
Brand: Scotsman™ AF80
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Extrudes ice flakes at a temperature just below zero degrees. Scotsman™ Ice Flaker with Storage bin can produce up to 70kg of flake per day and holds up to 25kg of storage. Flakes contain 25% residual water content and is stored inside an insulated, plastic lined containment bin with front opening for easy access.
Ice flakes are extruded at a temperature just below zero degrees and contain 25% residual water content. The flake ice is stored inside an insulated, plastic lined storage bin with front opening for easy access. Electrical supply 230V/50Hz.
  • Up to 70kg (154 lbs.) of flake ice production per day
  • Up to 25kg (55 lbs.) built-in ice storage
  • Pressure-based water level monitoring system may work with demineralised water as well as with standard tap water
  • Electronically controlled operation, with self-diagnostic functions with external alarm lights providing operational information for early alert and fast diagnosis of operating issues
  • Plug-n-play configuration right out of the box - just connect incoming water line and water discharge
  • External components constructed from corrosion-resistant stainless steel, crafted for optimal aesthetic appeal through superior fit and finish
  • Ergonomically convenient, slanted & disappearing storage bin door
  • Front access for easier maintenance
  • Front-in, front-out airflow allows for built-in installations
  • CFC-free R-134a refrigerant
  • 120mm legs included, extensions available as an option Scotsman flaked ice is ideal for a variety of applications within laboratory and industrial environments.
Specifications
Certifications/Compliance ISO 9001
Capacity (Metric) 25 kg
Depth (Metric) 626mm
Product Type AF80 Ice maker
For Use With (Application) May work with demineralised water as well as with standard water
Includes 120mm legs
Hertz 50Hz
Material Corrosion-resistant stainless steel
Height (Metric) 813mm
Voltage 230V
Width (Metric) 535mm
Yield 70kg/day
Electrical Requirements 230V/50Hz
Warranty 2 year manufacturer warranty