Learn More
Manganese(IV) Oxide Powder, MilliporeSigma™
Product Code. 15814720
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15814720 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15814720 Supplier Supelco Supplier No. 1059571000
 Restricted Item
Estimated Shipment: 19-11-2026
EMPLURA™ grade manganese(IV) oxide powder for laboratory applications.
TRUSTED_SUSTAINABILITY
Specifications
CAS 1313-13-9
Physical Form Powder
Quantity 1 kg
Molecular Formula MnO2
MDL Number MFCD00003463
UN Number UN 1479
Synonym manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein
InChI Key NUJOXMJBOLGQSY-UHFFFAOYSA-N
SMILES O=[Mn]=O
IUPAC Name dioxomanganese
Molecular Weight (g/mol) 86.94
PubChem CID 14801
Formula Weight 86.94
Percent Purity 89% (manganometric)
Odor Odorless
Packaging Glass bottle
Appearance Dark gray powder
Chemical Name or Material Manganese(IV) oxide