Learn More
MAN2B2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 17697-1-AP-150UL
 View more versions of this product
Product Code. 16847634
  • 4836.10 NOK / 150µL
Specifications
MAN2B2
Polyclonal
Unconjugated
MAN2B2
KIAA0935, MAN2B2
Rabbit
Antigen Affinity Chromatography
RUO
17160, 23324, 360955
-20°C
Liquid
Western Blot
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O54782, Q9Y2E5
Man2b2
MAN2B2 Fusion Protein Ag11952
150 μL
Primary
Human, Mouse, Rat