Learn More
Emma Safety Footwear Mammoet M1807 XD Safety Shoes
Product Code. 15999021
Change view
Click to view available options
Size:
37
38
39
40
41
42
43
44
45
46
47
48
49
Unit Size:
1 pair
Product Code. Size unitSize
15999021 37 1 pair
15909031 38 1 pair
15919031 39 1 pair
15929031 40 1 pair
15939031 41 1 pair
15949031 42 1 pair
15959031 43 1 pair
15969031 44 1 pair
15979031 45 1 pair
15989031 46 1 pair
15999031 47 1 pair
15909041 48 1 pair
15919041 49 1 pair
13 options
Product Code. 15999021
Brand: Emma Safety Footwear 76586837
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Extra high boot is heat and cold insulated. Ankle protection Comfort Sponge with added padding provides a more comfortable feel and better fit. Mammoet M1807 is very easy to put on and take off; the vertical eyelets on the shoe make lacing the fire-retardant laces very simple.
  • S3-Safety shoe with sole plate for wet work
  • GripForce™ Duo Heat Resistant Outsole
  • Heat and cold insulated
  • Fire-retardant laces
  • Waterproof Oil Nubuck construction
  • Hydro-Tec™ Sanitized Silver
Specifications
Certifications/Compliance EN ISO 20345:2011
Closure Type Laces
Color Black
Design Mammoet M1807 XD
Liner Material Hydro-Tec™ Sanitized Silver / Sympatex™
Material Oiled Nubuck
Size 37
Toe Material Steel