Learn More
Lonza™ Mammary Epithelial Growth Medium Bulletkit
Product Code. 11665430
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
Product Code. 11665430
Brand: Lonza™ CC3150
Optimized for the growth of Mammary Epithelial cells in a serum-free environment. Mammary Epithelial Growth Medium Bulletkit is a readily available media kit giving researchers the confidence and the convenience of a system that works to support their cells.
  • Referenced in over 130 unique publications primarily focusing on breast cancer research
  • Used for Lonza's primary HMECs as well as normal breast cell lines such as MCF-10A and MCF-12
  • Working with the same media between cell lines and primary cells can reduce variability within the experiments and simplifies the shift for researchers
  • In addition, if primary HMECs are utilized as a side-by-side control with normal breast cell lines, the MEGMTM BulletKitTM media may also offer cost-benefits since customers can use the same media to support both cell types
Order Info
dimProductType
TRUSTED_SUSTAINABILITY
Specifications
Product Type Growth Medium
Quantity 1 Kit
Content And Storage Kit which contains a 500mLl bottle of MEBM, (CC- 3151-mammary epithelial basal medium (no growth factors) (500 ml) ) and MEGM singlequots (CC-4136-supplements and growth factors (BPE, hydrocortisone, hEGF, insulin and gentamicin/ amphotericin-B)