Få mere at vide
IBA Lifesciences pCSG-IBA Mammalian Expression Vector with No Secretion
GREENER_CHOICE
Artikelnummer. 15623878 Shop alle IBA Lifesciences produkter
Klik for at se tilgængelige muligheder
Vector:
pCSG-IBA103
pCSG-IBA105
pCSG-IBA123
pCSG-IBA143
pCSG-IBA145
pCSG-IBA162
pCSG-IBA164
pCSG-IBA167
pCSG-IBA168
pCSG-IBA23
pCSG-IBA3
pCSG-IBA43
pCSG-IBA45
pCSG-IBA5
pCSG-IBA62
pCSG-IBA64
pCSG-IBAwt1
Pakningsstørrelse:
5µg
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15623878
Brand: IBA Lifesciences 55162001
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
Denne vare kan ikke returneres. Se returpolicy
Designed for high-level episomal expression in mammalian hosts under G418 selection. IBA Lifesciences GmbH Mammalian Expression Vector with No Secretion is a large expression vector with universal features for transient expression as well as for generation of stable cell lines (NeoR).
pCSG-IBA mammalian expression vectors
  • Designed for high-level episomal expression in mammalian hosts under G418 selection containing the following elements:
    • Human cytomegalovirus (CMV) immediate-early promoter for high-level expression in a wide range of mammalian cells
    • Episomal replication through Epstein Barr Virus replication origin (oriP) and nuclear antigen encoded by EBNA-1 in human, primate and canine cells and through SV40 replication origin in cells latently infected with SV40 or that express the SV40 large T antigen (e.g. HEK293T, COS-1, COS-7)
  • Stable cell lines can be selected by the neomycin resistance gene (NeoR).
  • In addition, the human cytomegalovirus (CMV) immediate-early promoter enables a high-level expression in a wide range of mammalian cells.
Please note that cloning into pCSG-IBA Acceptor Vectors compulsorily requires the restriction enzyme Esp3I since no other MCS for the integration of a gene of interest is available. Besides to the direct cloning of the gene of interest into pCSG-IBA vectors with Esp3I, another option via a so-called entry-vector is possible.
Tekniske data
Constitutive or Inducible System Constitutive
Marker Gene Ampicillin
Promoter CMV
Product Type Vector
Selection Agent (Eukaryotic) Geneticin™ (G-418)
Content And Storage Recommended: 2°C to 8°C for frequent usage, -20°C for long-term storage
Species Mammalian
Protein Tag Twin-Strep-tag, FLAG-tag
Cloning Method Direct Cloning
Concentration 250 ng/μL
Primer Forward
Quantity 5 μg
Vector pCSG-IBA162
For Use With (Application) Episomal expression (oriP, EBNA-1)
Vis mere Vis mindre
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.