missing translation for 'onlineSavingsMsg'
Learn More

Maltose Monohydrate, Millipore™

Product Code. 10332211
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10332211 25 g 25g
10026540 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10332211 Supplier Millipore Supplier No. 1059120025
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For biochemistry

for biochemistry

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6363-53-7

Specifications

Melting Point 125°C (anhydrous)
MDL Number MFCD00149343
Quantity 25 g
Synonym Maltose monohydrate, Maltose monohydrate
Molecular Weight (g/mol) 360.32
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.