missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Scharlab Malt Extract Agar
Click to view available options
:
Malt Extract Agar; Color: Yellow; Diameter: 90 mm
Description
- BAM
- Solid culture medium for the isolation and enumeration of fungi, with mycological peptone.
Specifications
Specifications
| Color | Yellow |
| Form | Viscous |
| Media Type | Malt Extract Agar |
| Product Type | Prepared Media Plates |
| Formulation | Malt Extract 30.0 g/L, Soy Peptone 3.0 g/L, Agar 15.0 g/L |
| For Use With (Application) | Isolation and Enumeration of Fungi |
| Incubation Conditions | 22.5°C ± 2°C |
| Incubation Time | 3 to 5 Days |
| Quantity | 20 Plates |
| Diameter (Metric) | 90 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction