Learn More
Malonyl dichloride, 94%
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Description
Malonyl Dichloride is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Malonyl Dichloride is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products.
Solubility
Decomposes in water.
Notes
Recommended storage temperature: 2 - 4°C. Incompatible with oxidizing agents.
Malonyl Dichloride is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products.
Solubility
Decomposes in water.
Notes
Recommended storage temperature: 2 - 4°C. Incompatible with oxidizing agents.
Specifications
Specifications
| Density | 1.449 |
| Boiling Point | 53°C to 55°C (19 mmHg) |
| Flash Point | 47°C (117°F) |
| Linear Formula | CH2(COCl)2 |
| Refractive Index | 1.465 |
| Quantity | 25 g |
| UN Number | UN2920 |
| Beilstein | 774044 |
| Solubility Information | Decomposes in water. |
| Formula Weight | 140.95 |
| Show More |