Learn More
Malondialdehyde Bis(dimethyl Acetal), Sigma-Aldrich™
Specifications
Specifications
| Density | 0.99 g/cm3 at 20°C |
| Boiling Point | 183°C |
| Molecular Formula | (CH3O)2CHCH2CH(OCH3)2 |
| MDL Number | MFCD00008488 |
| Quantity | 250 mL |
| Synonym | Malondialdehyde bis(dimethyl acetal), 1,1,3,3-Tetramethoxypropane, Malondialdehyde tetramethyl acetal, Malondialdehyde bis(dimethyl acetal) |
| Molecular Weight (g/mol) | 164.2 |
| Grade | Research |