Learn More
Malonaldehyde bis(diethyl acetal), 97%, Thermo Scientific Chemicals
Catalog Number p-3758288
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
122-31-6
C11H24O4
220.31
MFCD00009240
KVJHGPAAOUGYJX-UHFFFAOYSA-N
malonaldehyde bis diethyl acetal, tetraethoxypropane, malonaldehyde tetraethyl acetal, malonaldehyde diethyl acetal, usaf kf-26, propane, 1,1,3,3-tetraethoxy, tetraethyl malondialdehyde acetal, tetraethoxy propane, malonaldehyde tetraethyl diacetal, malonaldehyde, bis diethyl acetal
67147
1,1,3,3-tetraethoxypropane
CCOC(CC(OCC)OCC)OCC
122-31-6
C11H24O4
220.31
MFCD00009240
KVJHGPAAOUGYJX-UHFFFAOYSA-N
malonaldehyde bis diethyl acetal, tetraethoxypropane, malonaldehyde tetraethyl acetal, malonaldehyde diethyl acetal, usaf kf-26, propane, 1,1,3,3-tetraethoxy, tetraethyl malondialdehyde acetal, tetraethoxy propane, malonaldehyde tetraethyl diacetal, malonaldehyde, bis diethyl acetal
67147
1,1,3,3-tetraethoxypropane
CCOC(CC(OCC)OCC)OCC
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point -90.0°C
Color Colorless to Yellow
Density 0.9100g/mL
Boiling Point 220.0°C
Flash Point 88°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (C2H5O)2CHCH2CH(OC2H5)2
Refractive Index 1.4100 to 1.4120
Quantity 5 mL
Beilstein 01, III, 3095
Fieser 02,257
Specific Gravity 0.91
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,ether
Formula Weight 220.31
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Malonaldehyde bis(diethyl acetal)
Show More Show Less
compliance-icons
Product Identifier
  • Malonaldehyde bis(diethyl acetal)
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.