Learn More
Maleic Anhydride, TRC
Specifications
Specifications
| Chemical Name or Material | Maleic anhydride |
| CAS | 108-31-6 |
| Molecular Formula | C4 H2 O3 |
| Quantity | 10 g |
| Synonym | 2,5-Furandione, Maleic anhydride (8CI), 2,5-Dihydrofuran-2,5-dione, A 1018, BM 10, Baolilong A 1018, Crystal Man, Crystal Man AB, Dihydro-2,5-dioxofuran, GR 216, HA 59, HG 3-993-76, M 116389, M 188, Maleic acid anhydride, NSC 137651, NSC 137652, NSC 137653, NSC 9568, Nourymix MA 901, SY-A, Toxilic anhydride, WS 8201, cis-Butenedioic anhydride |
| InChI Formula | InChI=1S/C4H2O3/c5-3-1-2-4(6)7-3/h1-2H |
| SMILES | O=C1OC(=O)C=C1 |
| IUPAC Name | furan-2,5-dione |
| Molecular Weight (g/mol) | 98.06 |
| Formula Weight | 98.0004 |
Product Title
Ser du en mulighed for forbedring?