Learn More
Maleic Acid, Sigma-Aldrich™
Product Code. 15898240
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
50 kg
Unit Size:
100g
1kg
500g
50kg
5kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15898240 5 kg 5kg
15878240 100 g 100g
15846230 500 g 500g
15888240 1 kg 1kg
15808250 50 kg 50kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 15898240 Supplier Sigma-Aldrich Supplier No. 8003805000
 Restricted Item
Estimated Shipment: 23-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 110-16-7
Specifications
Melting Point 130°C to 135°C (lit.)
Density 1.59 g/mL at 25°C
Quantity 5 kg
Molecular Formula HO2CCH=CHCO2H
MDL Number MFCD00063177
Synonym cis-Butenedioic acid, Toxilic acid, cis-Butenedioic acid, Toxilic acid
Molecular Weight (g/mol) 116.07
Grade Ungraded
Chemical Name or Material Maleic Acid