Learn More
Idex Male-Threaded Nuts
Product Code. p-66663
Change view
Click to view available options
For Use With (Equipment):
1/32 in. or 1/16 in. OD Tubing
1/8 in. O.D. Tubing
Length (English):
0.87 in.
Material:
PEEK
Width (English):
0.365 in.
Unit Size:
Each
Pack of 10
Product Code. For Use With (Equipment) Length (English) Material Width (English) unitSize
12977589 1/32 in. or 1/16 in. OD Tubing 0.87 in. PEEK 0.365 in. Pack of 10
12917629 1/8 in. O.D. Tubing 0.87 in. PEEK 0.365 in. Each
2 options
Product Code. 12977589
Brand: Idex 0201373
Only null left
Add to basket
Add to basket
Combine with Super Flangeless™ ferrule for a high pressure holding flat-bottom fitting assembly
  • Eliminates tubing twist that can loosen connections
  • Resistant to loosening through vibration
Specifications
Type Super Flangeless Nut
Color Natural
Diameter (English) Outside 0.0625 in.
Length (English) 0.87 in.
Width (English) 0.365 in.
For Use With (Equipment) 1/32 in. or 1/16 in. OD Tubing
Material PEEK