Learn More
Thermo Scientific™ Male Luer-LOK Priming Syringes
Product Code. 10409254 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
1 mL
2.5 mL
5 mL
10 mL
25 mL
50 mL
Product Type:
GC Autosampler Priming Syringe, Luer-Lok
GC Autosampler Syringe
Gas Tight GC Autosampler Syringe, Luer-Lok
Unit Size:
Each
Product Code. Volume (Metric) Product Type unitSize
10409254 2.5 mL Gas Tight GC Autosampler Syringe, Luer-Lok Each
10524655 1 mL Gas Tight GC Autosampler Syringe, Luer-Lok Each
13436886 10 mL GC Autosampler Syringe Each
10137854 5 mL Gas Tight GC Autosampler Syringe, Luer-Lok Each
10585995 10 mL Gas Tight GC Autosampler Syringe, Luer-Lok Each
10409444 25 mL Gas Tight GC Autosampler Syringe, Luer-Lok Each
10351305 50 mL GC Autosampler Priming Syringe, Luer-Lok Each
7 options
Product Code. 10409254
Brand: Thermo Scientific™ 365LL541
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Thermo Scientific™ Male Luer-LOK Priming Syringes offer easy, accurate and reproducible manual injections.
Thermo Scientific™ Male Luer-LOK Priming Syringes offer easy, accurate and reproducible manual injections.
  • Square tip to prevent damage to the injector
  • Wide range of volumes
  • Precision made from borosilicate glass and stainless steel
  • Robust design and easy-to-read markings
TRUSTED_SUSTAINABILITY
Specifications
Product Type Gas Tight GC Autosampler Syringe, Luer-Lok
Volume (Metric) 2.5 mL
For Use With (Equipment) Gas Chromatography Systems
Replacement Needle 365RNL22