Description
Thermo Scientific™ Male Luer-LOK Priming Syringes offer easy, accurate and reproducible manual injections.
- Square tip to prevent damage to the injector
- Wide range of volumes
- Precision made from borosilicate glass and stainless steel
- Robust design and easy-to-read markings
Specifications
Specifications
| Product Type | Gas Tight GC Autosampler Syringe, Luer-Lok |
| Volume (Metric) | 5 mL |
| For Use With (Equipment) | Gas Chromatography Systems |
| Replacement Needle | 365RNL22 |