Description
Luer fittings allow for a simple yet secure twist-lock connection. Ideal for capping off fluid transfer system. Prevent leaks with integral lock ring mechanism
Specifications
Specifications
| Certifications/Compliance | Non-DEHP;Non-Phthalate;RoHS |
| Inlet | Male Luer Integral Lock Ring, 1/4-28 UNF(M) Thread |
| Material (Seal) | Nylon |
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.