Description
Luer fittings allow for a simple yet secure twist-lock connection. Ideal for capping off fluid transfer system. Prevent leaks with integral lock ring mechanism
Specifications
Specifications
| Certifications/Compliance | ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP |
| Inlet | Male Luer Integral Lock Ring, 10-32(M) Special Tapered Thread |
| Material (Seal) | Polypropylene |
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.