missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAK16 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17505-1-AP-150UL
This item is not returnable.
View return policy
Description
MAK16 gene ontology annotations related to this gene include RNA binding.Specifications
| MAK16 | |
| Polyclonal | |
| Unconjugated | |
| MAK16 | |
| MAK16, MAK16 homolog (S. cerevisiae), MAK16L, NNP78, Protein MAK16 homolog, Protein RBM13, RBM13 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 306526, 67920, 84549 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8BGS0, Q9BXY0 | |
| MAK16 | |
| MAK16 Fusion Protein Ag11705 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction