Learn More
3M™ Maintenance Sorbent Roll
Description
- Designed for use in and around the work area to catch leaks and drips
- For use on most non-aggressive liquids
- Very easy to use and to retrieve
- Designed to cover large surfaces
Specifications
Specifications
| Color | Gray |
| Capacity (English) | 34.8 gal. |
| Capacity (Metric) | 131.7 L |
| Type | Sorbent Roll |
| For Use With (Application) | Workplace safety |
| Model | M-B2001 |
| Dimensions (L x W) | 45.7 x 0.40 m |
| Length (English) | 150 ft. |
| Length (Metric) | 45.7 m |
| Width (English) | 15.5 in. |
| Show More |