Learn More
Magnolol, MedChemExpress
Product Code. 16424777
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. Quantity unitSize
16424777 50 mg 50mg
16419016 100 mg 100mg
16418888 10 mg 10mg
3 options
Product Code. 16424777
Brand: Medchem Express HYN0163
MedChemExpress Magnolol, a natural lignan isolated from the stem bark of Magnolia officinalis, is a dual agonist of both RXRα and PPARγ, with EC50 values of 10.4 μM and 17.7 μM, respectively.
Magnolol, a natural lignan isolated from the stem bark of Magnolia officinalis, is a dual agonist of both RXRα and PPARγ, with EC50 values of 10.4 μM and 17.7 μM, respectively.
Specifications
Chemical Name or Material Magnolol
CAS 528-43-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C18H18O2
Quantity 50 mg
Solubility Information DMSO : 100 mg/mL (375.47 mM; Need ultrasonic)
SMILES OC1=CC=C(CC=C)C=C1C2=CC(CC=C)=CC=C2O
Molecular Weight (g/mol) 266.33
Formula Weight 266.33
Percent Purity 99.92%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death