missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MagniSort™ Human CD56 Positive Selection Kit
Description
Includes
- MagniSort Anti-Human CD56 Biotin : 200 tests, 20μL/test
- MagniSort Positive Selection Beads A
- CD56+ cells are bound by antibody and then magnetic beads
- When placed in a magnetic field, the undesired cells can be separated from CD56+ cells by decanting
- After positive selection, the purity of selected cells can be verified by staining with Anti-Human CD56 (clone CMSSB)
Specifications
Specifications
| Cell Type | CD56+ |
| Product Type | Positive Selection Kit |
| No. of Tests | 200 tests |
| Quantity | 200 tests |
| Target Species | Human |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?