Learn More
3M™ Magnifying Lens
Product Code. 15879644 Shop All 3M Products
Change view
Click to view available options
Power Intensity:
2.0X
2.5X
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Power Intensity Unit Size
15879644 2.5X Each
15869634 2.0X Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15879644 Supplier 3M™ Supplier No. 7000144517
  • ONLINE EXCLUSIVE PRICE
    400.00 NOK
    290.91 NOK / Pack of 1
    Save 109.09 NOK
    27% Off
Estimated Shipment: 24-02-2027
3M™ Magnifying Lens is used for magnifying object and can be used with all Speedglas welding helmets.
  • On the 3M™ Speedglas™ Welding Helmet Series 9100 there is a magnifying lens holder built into the welding filter, making lens installation easier and more secure
  • For the new 3M™ Speedglas™ Welding Helmet Series 100 there is a magnifying lens holder built into the welding filter, making lens installation easier and more secure. For the older 100 series filters you will need a holder for the magnifier
  • For the 3M™ Speedglas™ Welding Helmet SL you will need to add the holder for magnifier as a separate spare part (171031)
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Speedglas
Power Intensity 2.5X
Product Type Magnifying Lens