Description
- Opaque PTFE construction
- High performance permanent magnets
- Long lifespan
- Exceptional temperature resistance
- Wide range of sizes
- Stirring bars with diameters of 6 and 8mm can be used with Buerkle™ Fish-clip™ for extra gentle stirring
Specifications
Specifications
| Color | White |
| Diameter (Metric) | 7 mm |
| Length (Metric) | 30 mm |
| Shape | Cylinder |