Learn More
Velp Scientifica™ Magnetic Stir Bars
Product Code. 15875221 Shop All Velp Products
Change view
Click to view available options
Length (Metric):
5 mm
6 mm
8 mm
10 mm
13 mm
30 mm
Diameter (Metric):
8 mm
10 mm
20 mm
Unit Size:
Each
Product Code. Length (Metric) Diameter (Metric) unitSize
15875221 6 mm 10 mm Each
15805231 13 mm 10 mm Each
15845211 5 mm 10 mm Each
15865221 8 mm 20 mm Each
15885221 10 mm 20 mm Each
15895221 30 mm 8 mm Each
6 options
Product Code. 15875221
Brand: Velp Scientifica™ A00000354
Only null left
Add to basket
Add to basket
Velp Scientifica™ Magnetic Stir Bars for use with Velp Scientifica™ Hotplate Stirrers.
  • For use with Velp Hotplate Stirrers
  • Available in various sizes and shapes
Specifications
For Use With (Equipment) Hotplate Stirrers
Diameter (Metric) 10 mm
Length (Metric) 6 mm
Shape Disc