Learn More
Buerkle™ Magnetic Centrifugal Pumps
Shop All Bürkle Products
Click to view available options
Flow Rate:
1 to 24 L/min.
Conditionnement:
Each
Description
The drive, via contact-free magnetic force transfer, requires no dynamic shaft seals and this makes a hermetically sealed pump housing possible,which means that it cannot leak.
- Seal-less
- Surrounding temperature max. 45 °C
- Operating temperature max. 60 °C
- Viscosity max. 20 mPas
Spécification
Spécification
| Model | Pressure connection 3/4 in. outer thread (BSP) |
| Voltage | 230 V |
| Flow Rate | 1 to 24 L/min. |
| Applications | For delivery heights up to 3 m |
| Temperature (Metric) Max | 60°C Operating temperature |
| Drive Unit | 29 Watts |
| Material | Polypropylene (Fiber Glass Reinforced), Stainless Steel (Shaft), Rulon (Bearing), FKM (Seal), Ceramics (Shaft) |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu