Learn More
Magnesiumacetylacetonate, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 14024-56-7 |
| Molecular Formula | C10H14MgO4 |
| MDL Number | MFCD00013497 |
| Quantity | 5 g |
| SMILES | CC([CH-]C(C)=O1)=O[Mg+2]12O=C([CH-]C(C)=O2)C |
| Molecular Weight (g/mol) | 222.52 |
| Percent Purity | 98% |
| Chemical Name or Material | Magnesiumacetylacetonate |
Product Title
Haben Sie Verbesserungsvorschläge?