Learn More
Hanna Instruments™ Ammonia Reagent Sets, For Colorimetric Water Testing
Product Code. 10102181
Click to view available options
For Use With (Application):
Ammonia High Range
Magnesium and Total Hardness
No. of Tests:
25
300
Unit Size:
Each
Product Code. 10102181
Brand: Hanna Instruments™ HI9371903
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Allows users to achieve fast and accurate colorimetric measurements. Hanna Instruments™ Ammonia Reagent Sets, For Colorimetric Water Testing are high-quality, pre-measured solutions for use with compatible benchtop and/or portable Hanna meters.
These high quality reagents are designed to follow accepted test methods for colorimetric determination of various parameters of water hardness. By simply adding the reagents to the sample as directed, a reaction will take place and a compatible meter can be used to determine the concentration from the color that is produced.
  • Reagent sets are available for determination of:
    • Calcium Hardness (using liquid or powdered reagents)
    • Calcium and Magnesium Hardness (high range)
    • Magnesium and Total Hardness
    • Total Hardness (low, medium and/or high range)
  • Pre-made, pre-measured water testing reagents for ease of use
  • Prepared with high purity chemicals manufactured in Hanna's state-of-the-art facility
  • Clearly marked with expiration date and lot number on each packet and bottle for traceability
Specifications
For Use With (Application) Magnesium and Total Hardness
For Use With (Equipment) Compatible Hanna benchtop and portable photometers
Includes Dropper bottles
No. of Tests 300
Quantity 1 Set
Range 0 to 2.00 mg/L (Mg Hardness), 0 to 4.70 mg/L (Total Hardness)
Test Method Calmagite Colorimetric
Type Water Testing Kit
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Reagents for 300 tests magnesium and hardness .
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H373-May cause damage to organs.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P390-Absorb spillage to prevent material damage.
Supplemental information
  • MIXTURE LIST-Contains : Sodium hydroxide, Tetrasodium EDTA