Learn More
WTW™ Magnesium Cell Test Kits
Description
- Convenient and cost-effective
- Assured quality by AQA/IQC
- For infrequent or very large sample throughput in the laboratory
- Sample adding, inserting, measuring and reading
- Reaction cuvette with barcode or method selection
Specifications
Specifications
| Analytes | Iron(III) thiocyanate |
| For Use With (Application) | For drinking water, surface water |
| For Use With (Equipment) | PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral, Photoflex photometers |
| Includes | Certificates |
| No. of Tests | 25 |
| Quantity | 1 Pack |
| Range | 5.0 - 75.0mg/L |
| Test Method | Photometric |
| Type | Chlorine test kit |