missing translation for 'onlineSavingsMsg'
Learn More

Magnesium sulfate anhydrous, 98.0%, for analysis EMSURE™, MilliporeSigma™

Product Code. 15824770
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15824770 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15824770 Supplier Supelco Supplier No. 1060671000
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 1124°C
CAS 7487-88-9
pH 7.9 (50 g/L, H2O, 25°C)
Quantity 1 kg
Assay Percent Range ≥ 98.0%
Molecular Formula MgO4S
Solubility Information 300 g/L
InChI Key CSNNHWWHGAXBCP-UHFFFAOYSA-L
SMILES [Mg++].[O-]S([O-])(=O)=O
IUPAC Name magnesium(2+) sulfate
Molecular Weight (g/mol) 120.36
Packaging Glass Bottle
Density 2.66 g/cm3 (at 20°C); Bulk Density: 600 kg/m3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.