Learn More
Magnesium Matrix Modifier, Supelco™
Specifications
Specifications
| Density | 1.05 g/cm3 at 20°C |
| pH | <1 (20°C in H2O) |
| Type | Magnesium Modifier |
| Quantity | 50 mL |
| Grade | Research |
| Chemical Name or Material | Magnesium Matrix Modifier |